Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray AUTION EYE™ Control Solution H/L
SDP

Catalog No. 21403498
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-403-498 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: For in vitro diagnostic use only. Handle according to laboratory safety guidelines. Avoid contact with skin or eyes. Store as directed and use protective equipment when handling.
Catalog No. 21-403-498 Supplier Arkray Supplier No. 108157
Add to Cart
Add to Cart
Warning: For in vitro diagnostic use only. Handle according to laboratory safety guidelines. Avoid contact with skin or eyes. Store as directed and use protective equipment when handling.

This Solution provides reliable high and low-level quality control for A1c analyzers, ensuring accurate, consistent results and supporting efficient lab workflow with minimal maintenance.

  • High and Low-Level Control: Provides both high and low control levels to verify analyzer accuracy across clinically relevant ranges.
  • Reliable Quality Control: Ensures precise, consistent performance of A1c analyzers, helping maintain trustworthy lab results.
  • Ready-to-Use Solution: Minimizes preparation time and reduces potential handling errors, streamlining daily QC workflow.
  • Compatibility: Designed specifically for use with Arkray A1c analyzers to optimize system performance.
  • Supports Regulatory Compliance: Helps laboratories meet quality control standards and regulatory requirements.
  • Stable and Consistent: Formulated for extended shelf life and batch-to-batch consistency, reducing the need for frequent replacement.
  • Time-Saving: Reduces downtime with easy integration into daily lab routines, improving overall la

Specifications

Sample Type Urine
Type Control Solution
Disposable Single-use
Sterility Sterile
Form Liquid

AUTION EYE Control Solution H/L ensures accurate high and low-level quality control for Arkray A1c analyzers, supporting consistent results and efficient lab workflow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.