Learn More
Arkray Aution Sticks 10EA
SDP
Catalog No. 21381516
Change view
Click to view available options
Quantity:
100 Strips/Bottle
Catalog No. Quantity
21-381-516 100 Strips/Bottle
1 options
Exact match for part number 73526
Catalog No. 21-381-516 Supplier Arkray Supplier No. 73526
Add to Cart
Add to Cart
Exact match for part number 73526
Aution Sticks 10EA urine test strips provide reliable, multi-parameter urinalysis testing for efficient clinical screening and routine laboratory workflows.
  • Compatible with select Arkray urine analyzers for streamlined urinalysis workflows
  • Provides efficient screening support for routine patient testing and health monitoring
  • Delivers consistent and standardized test performance for dependable results
  • Convenient strip format allows for simple handling and easy operation
  • Supports improved laboratory productivity through fast and efficient processing
  • Ideal for hospitals, physician office laboratories, clinics, and diagnostic facilities
  • Manufactured to support quality urinalysis testing and workflow consistency
  • For in vitro diagnostic use only. Use according to instructions for compatible Arkray analyzers to ensure accurate and reliable urinalysis results.
Specifications
CE Marker Self-declared
Clia Complexity Waived (Urine)
Detectable Analytes Glucose, Protein, Bilirubin, pH, Ketone
Type Urine Test Strips
Quantity 100 Strips/Bottle
Form Ready-to-Use (RTU)