Learn More
Arkray AUTION Sticks, 9EB
SDP
Catalog No. 21403502
Change view
Click to view available options
Quantity:
100 Strips/Bottle
Catalog No. Quantity
21-403-502 100 Strips/Bottle
1 options
Warning: For in vitro diagnostic use only. Do not ingest. Avoid contact with eyes and skin. Keep out of reach of children. Store as directed and use within expiration date. Handle according to laboratory safety guidelines.
Catalog No. 21-403-502 Supplier Arkray Supplier No. 110734
Add to Cart
Add to Cart
Warning: For in vitro diagnostic use only. Do not ingest. Avoid contact with eyes and skin. Keep out of reach of children. Store as directed and use within expiration date. Handle according to laboratory safety guidelines.
AUTION Sticks, 9EB are rapid urine test strips designed for reliable multi-parameter screening. They provide quick, easy-to-read results for clinical labs and point-of-care settings, supporting efficient urinalysis with consistent performance.
  • 9 Parameter Test Strip: Provides multi-parameter urine screening in a single strip for efficient and comprehensive analysis.
  • Fast Results: Delivers rapid colorimetric results to support quick decision-making and efficient workflow.
  • Easy to Use: Simple dip-and-read format designed for both clinical labs and point-of-care testing environments.
  • Accurate Screening: Supports reliable urine chemistry screening with consistent performance and clear results.
  • Reduced Manual Work: Helps reduce manual testing steps and supports faster throughput for high-volume testing.
  • Compact and Convenient: Portable test strips are easy to store and handle, supporting flexible testing workfl
Specifications
CE Marker CE-IVD
Clia Complexity High, Moderate
Type Test Strips
Disposable Single-use
Quantity 100 Strips/Bottle
Form Solid
AUTION Sticks, 9EB provide quick, reliable multi-parameter urine screening for clinical and point-of-care settings, supporting efficient workflow and accurate urinalysis results.