Learn More
Electron Microscopy Sciences Auto Desiccator Cabinet, Horizontal, 120 VAC
SDP
Catalog No. 501899509
Encompass_Preferred
Made from clear methyl-methacrylate (acrylic), this auto-desiccator cabinet features a permanent desiccant contained in the control unit so no desiccant tray is needed. The cycle is indicated by a green light for a 4 1/2 hours absorption period, or a red light indicating 1/2 hour regeneration of the desiccant. This cabinet will lower relative humidity to 30 - 40% and it includes two adjustable shelves. The unit draws 6 watts, 120 VAC. Horizontal layout.
: 24½" x 15½"
: 13½"
Catalog No. 50-189-9509 Supplier Electron Microscopy Sciences Supplier No. 71217HZ
Add to Cart
Add to Cart
Made from clear methyl-methacrylate (acrylic), this auto-desiccator cabinet features a permanent desiccant contained in the control unit so no desiccant tray is needed. The cycle is indicated by a green light for a 4 1/2 hours absorption period, or a red light indicating 1/2 hour regeneration of the desiccant. This cabinet will lower relative humidity to 30 - 40% and it includes two adjustable shelves. The unit draws 6 watts, 120 VAC. Horizontal layout.
: 24½" x 15½"
: 13½"