Learn More
Azzota Corp Auto Digital Abbe Refractomete
Catalog No. 5000887240
Encompass_Preferred
Automatic Digital Abbe Refractometer measurement range 1 3000-1 7000 nD 0-100% Brix LCD display
Catalog No. 50-008-87240 Supplier Azzota Corp Supplier No. AR4
Automatic Digital Abbe Refractometer measurement range 1 3000-1 7000 nD 0-100% Brix LCD display