Learn More
MSC ZEP Lime and Scale Remover, Liquid, Acidic, 12/Case
Catalog No. 502818943
Encompass
This lime and scale remover is an acidic liquid detergent designed for effective removal of mineral deposits and scale. It is a clear, colorless liquid with a mild odor, ideal for industrial and commercial cleaning applications.
  • Formulated as a liquid acidic detergent
  • Features a mild odor
  • Has a pH between 1 and 1.5
  • Each case contains twelve 32 oz. bottles
  • Classified as hazardous material
Catalog No. 50-281-8943 Supplier MSC Supplier No. 17596602
This lime and scale remover is an acidic liquid detergent designed for effective removal of mineral deposits and scale. It is a clear, colorless liquid with a mild odor, ideal for industrial and commercial cleaning applications.
  • Formulated as a liquid acidic detergent
  • Features a mild odor
  • Has a pH between 1 and 1.5
  • Each case contains twelve 32 oz. bottles
  • Classified as hazardous material