Learn More
MSC AUTO DISHWASH DETERGENT
Catalog No. 502819248
Encompass
Zep Provisions SMS HW Detergent is an odourless, liquid warewashing solution. It presents as a clear, orange-red liquid with a highly alkaline pH, making it effective for demanding cleaning applications.
  • Liquid form for easy dispensing
  • Clear, orange, red color for visual identification
  • Odourless formulation prevents fragrance contamination
  • High pH range of 12.5–13 ensures powerful cleaning
  • Completely soluble in water for efficient rinsing
  • Contains sodium metasilicate (disodium salt) as a key component
Catalog No. 50-281-9248 Supplier MSC Supplier No. 17595836
Zep Provisions SMS HW Detergent is an odourless, liquid warewashing solution. It presents as a clear, orange-red liquid with a highly alkaline pH, making it effective for demanding cleaning applications.
  • Liquid form for easy dispensing
  • Clear, orange, red color for visual identification
  • Odourless formulation prevents fragrance contamination
  • High pH range of 12.5–13 ensures powerful cleaning
  • Completely soluble in water for efficient rinsing
  • Contains sodium metasilicate (disodium salt) as a key component