Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Lime and Scale Remover, Liquid, Acidic, 1 gal, 4/Case

Catalog No. 502868836
Encompass

This product is a liquid, acidic cleaner formulated to effectively remove lime and scale. It is designed for industrial applications requiring a moderately harsh solution.

  • Effectively removes lime and scale
  • Liquid formula for easy application
  • Mild scent
  • Packaged in 1 gal bottles, 4 per case
  • Low pH of 1–1.5
  • Density of 1.259 g/cm3
  • Water soluble
  • Contains orthophosphoric acid

Catalog No. 50-286-8836 Supplier MSC Supplier No. 17596594
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a liquid, acidic cleaner formulated to effectively remove lime and scale. It is designed for industrial applications requiring a moderately harsh solution.

  • Effectively removes lime and scale
  • Liquid formula for easy application
  • Mild scent
  • Packaged in 1 gal bottles, 4 per case
  • Low pH of 1–1.5
  • Density of 1.259 g/cm3
  • Water soluble
  • Contains orthophosphoric acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.