Learn More
Grainger Speedaire Electric Auto Drain Valve, Brass, 3/8 in. NPT, 100-120 V, 1 ea
Catalog No. 502771993
Encompass_Preferred
This electric auto drain valve is designed for the automatic removal of condensate from pneumatic systems. Featuring a durable brass body, this unit provides a reliable drain rate of 4.33 GPM and operates within a voltage range of 100-120 V.
  • Brass construction for durability
  • 3/8 in. NPT pipe connection
  • Adjustable drain cycle from 0.5 sec. to 45 min.
  • Purge time adjustable from 0.5 sec. to 10 sec.
  • IP65 enclosure rating
  • Operating temperature range of 14°F to 140°F
  • Includes 6 ft. power cord
  • Max. pressure of 250 psi
Catalog No. 50-277-1993 Supplier Grainger Supplier No. 13X526
This electric auto drain valve is designed for the automatic removal of condensate from pneumatic systems. Featuring a durable brass body, this unit provides a reliable drain rate of 4.33 GPM and operates within a voltage range of 100-120 V.
  • Brass construction for durability
  • 3/8 in. NPT pipe connection
  • Adjustable drain cycle from 0.5 sec. to 45 min.
  • Purge time adjustable from 0.5 sec. to 10 sec.
  • IP65 enclosure rating
  • Operating temperature range of 14°F to 140°F
  • Includes 6 ft. power cord
  • Max. pressure of 250 psi