Learn More
Grainger Speedaire Electric Auto Drain Valve, Brass, 1/2 in. NPT, 16.5 GPM, 100 V to 120 V
Catalog No. 502771871
Encompass_Preferred
This electric auto drain valve is designed for efficient moisture removal in pneumatic systems. Constructed from durable brass, the valve operates within a voltage range of 100 V to 120 V and includes a 6 ft. power cord and filter ball valve for installation.
  • Adjustable drain cycle from 0.5 sec. to 45 min.
  • Purge time of 0.5 sec. to 10 sec.
  • 16.5 gpm flow rate
  • IP65 rated enclosure
  • Maximum pressure of 300 psi
  • Operating temperature range of 14°F to 140°F
  • 1/2 in. NPT pipe connection and 3/8 in. drain size
Catalog No. 50-277-1871 Supplier Grainger Supplier No. 13X532
This electric auto drain valve is designed for efficient moisture removal in pneumatic systems. Constructed from durable brass, the valve operates within a voltage range of 100 V to 120 V and includes a 6 ft. power cord and filter ball valve for installation.
  • Adjustable drain cycle from 0.5 sec. to 45 min.
  • Purge time of 0.5 sec. to 10 sec.
  • 16.5 gpm flow rate
  • IP65 rated enclosure
  • Maximum pressure of 300 psi
  • Operating temperature range of 14°F to 140°F
  • 1/2 in. NPT pipe connection and 3/8 in. drain size