Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Auto Drain Valve 115V 1/4 in 0

Supplier:  Grainger 13X536

Encompass_Preferred

Auto Drain Valve 115V 1/4 in 0.75 gpm

Catalog No. 50-277-0130


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.