Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America Auto Emergency Response Kit
SDP

Catalog No. 17985125
Change view
Click to view available options
Weight (English):
13 lb.
17 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
17-985-125 13 lb.
17-985-126 17 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-985-125 Supplier Ready America Supplier No. 70352
Only null left
Add to Cart
Add to Cart

Contains critical supplies for an automotive emergency

  • Our Auto Emergency kit is packed in duffle bag that contains critical supplies for an automotive emergency
  • The kit includes needed basic supplies such as jumper cables, first aid kit, distress banner, and tire fixer plus more
  • A very convenient carry bag with ample room so you can add more auto repair supplies so you can customize your kit

Specifications

Product Type Auto Response Emergency Kit
Dimensions (L x W x H) 16.25 x 10.25 x 8.5 in.
Includes First Aid Kit
Weight (English) 13 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.