Learn More
Euthanex Auto Flow Anesthesia Machine with New TEC 3 Vaporizer - Funnel Fill and (5) output circuits.
Catalog No. 501540173
Encompass_Preferred
Auto Flow Anesthesia Machine with New TEC 3 Vaporizer and Funnel Fill well. Provides precise, consistent, automatic preset flow rates from each outlet without the need for manual flow meter adjustments. Includes (4) breather circuits and (1) induction chamber circuit. Use with Isoflurane only.
Catalog No. 50-154-0173 Supplier Euthanex Supplier No. EZ190F
Add to Cart
Add to Cart
Request a Quote
Auto Flow Anesthesia Machine with New TEC 3 Vaporizer and Funnel Fill well. Provides precise, consistent, automatic preset flow rates from each outlet without the need for manual flow meter adjustments. Includes (4) breather circuits and (1) induction chamber circuit. Use with Isoflurane only.