Learn More
World Precision Instrument Auto Flow Vaporizer Unit
SDP
Catalog No. 502538839
Encompass
The vaporizer is a precision instrument that blends incoming oxygen with liquid isoflurane and outputs anesthetic gas at the set oxygen to isoflurane ratio. The dial setting allows fine gradations for precise isoflurane concentration. The vaporizer is available in three configurations that incorporate output to an anesthesia system or as a stand-alone unit. The Classic Anesthesia System (EZ-7000) and the Single Animal Anesthesia System (EZ-B800)  are also available without a vaporizer, allowing you to incorporate your own unit.
Catalog No. 50-253-8839 Supplier World Precision Instrument Supplier No. EZ190AF
Add to Cart
Add to Cart
Request a Quote
The vaporizer is a precision instrument that blends incoming oxygen with liquid isoflurane and outputs anesthetic gas at the set oxygen to isoflurane ratio. The dial setting allows fine gradations for precise isoflurane concentration. The vaporizer is available in three configurations that incorporate output to an anesthesia system or as a stand-alone unit. The Classic Anesthesia System (EZ-7000) and the Single Animal Anesthesia System (EZ-B800)  are also available without a vaporizer, allowing you to incorporate your own unit.