Learn More
Braintree Scientific Auto FlowVaporizer Unit w/Isoflurane Vaporizer
SDP
Catalog No. 501954784
Encompass
Auto Flow Vaporizer MachineAutomatic five output manifold that does not require flowmeters. Precise, consistent, preset flow rate from each outlet, no variables EZ-190F Auto Flow Vaporizer Machine Features: Fixed flow rates are set specifically for mice and rats Each output is individually controlled with simple on/off switch Five outputs supply anesthesia to the induction chamber and up to four breathers Preset flow rates eliminate need for flowmeters and manual adjustments Protocols are easily followed without calculations Minimum opportunity for operator errorFlow rate is not impacted by the type or number of accessories connected Precise, low flow gas saves money and protects personnel Pushbutton oxygen flush purges induction chamber before removing animals Carry handle facilitates safe, easy transport of the machine **Sold for research purposes only and not for human use**
Catalog No. 50-195-4784 Supplier Braintree Scientific Supplier No. EZ190AF
Add to Cart
Add to Cart
Request a Quote
Auto Flow Vaporizer MachineAutomatic five output manifold that does not require flowmeters. Precise, consistent, preset flow rate from each outlet, no variables EZ-190F Auto Flow Vaporizer Machine Features: Fixed flow rates are set specifically for mice and rats Each output is individually controlled with simple on/off switch Five outputs supply anesthesia to the induction chamber and up to four breathers Preset flow rates eliminate need for flowmeters and manual adjustments Protocols are easily followed without calculations Minimum opportunity for operator errorFlow rate is not impacted by the type or number of accessories connected Precise, low flow gas saves money and protects personnel Pushbutton oxygen flush purges induction chamber before removing animals Carry handle facilitates safe, easy transport of the machine **Sold for research purposes only and not for human use**