Learn More
MSC World Dryer Electric Hand Dryer, 950 W, Silver Finish, Surface-mounted, ADA Compliant
Catalog No. 502835528
Encompass
This electric hand dryer features a 950 W power rating and a silver finish, designed for surface-mounted installation. It is ADA compliant and offers efficient, hygienic hand drying for various environments.
  • Surface-mounted ADA compliant design
  • Simplifies installation and reduces costs
  • Up to 40% faster dry time compared to leading competitive surface-mounted dryers
  • Up to 70% less energy consumption than competitive dryers
  • Universal voltage input (110-120 / 208 / 220-240 VAC)
  • ON/OFF heat control for customized comfort and energy savings
  • Incorporates SteriTouch antimicrobial technology to inhibit bacteria, mold, and fungus growth
  • Automatic activation for touch-free operation
  • IP23 rating provides protection against harmful water ingress
  • Ultra compact, durable, and vandal resistant construction
Catalog No. 50-283-5528 Supplier MSC Supplier No. 54858691
This electric hand dryer features a 950 W power rating and a silver finish, designed for surface-mounted installation. It is ADA compliant and offers efficient, hygienic hand drying for various environments.
  • Surface-mounted ADA compliant design
  • Simplifies installation and reduces costs
  • Up to 40% faster dry time compared to leading competitive surface-mounted dryers
  • Up to 70% less energy consumption than competitive dryers
  • Universal voltage input (110-120 / 208 / 220-240 VAC)
  • ON/OFF heat control for customized comfort and energy savings
  • Incorporates SteriTouch antimicrobial technology to inhibit bacteria, mold, and fungus growth
  • Automatic activation for touch-free operation
  • IP23 rating provides protection against harmful water ingress
  • Ultra compact, durable, and vandal resistant construction