Learn More
VAZYME INTERNATIONAL LLC Vazyme Automatic Nucleic Acids Extraction Instrument, 96 Throughput, Magnetic Bead Technology
Catalog No. NC3990443
Encompass_Preferred
The Vazyme VaPura 96 is an automatic nucleic acids extraction instrument capable of 96-throughput. It utilizes magnetic bead technology to purify DNA, RNA, protein, and cells from various sample types, offering an automated, fast, and simple operation.
  • Automatic extraction and purification system for DNA/RNA, protein, and cells.
  • Utilizes magnetic beads for adsorption, transfer, and release by magnetic rods and sleeves.
  • Automated, fast, and simple operation.
  • Processes 1 to 96 samples simultaneously using a 96-well deep-well plate.
  • Also processes 1 to 24 samples simultaneously using a 24-well deep-well plate.
  • Compatible with various magnetic beads nucleic acid extraction reagents.
  • Extracts and purifies nucleic acids from human blood and body fluids.
  • Primarily used for the extraction and purification of nucleic acids in human samples.
Catalog No. NC3990443 Supplier VAZYME INTERNATIONAL LLC Supplier No. VNP96P
The Vazyme VaPura 96 is an automatic nucleic acids extraction instrument capable of 96-throughput. It utilizes magnetic bead technology to purify DNA, RNA, protein, and cells from various sample types, offering an automated, fast, and simple operation.
  • Automatic extraction and purification system for DNA/RNA, protein, and cells.
  • Utilizes magnetic beads for adsorption, transfer, and release by magnetic rods and sleeves.
  • Automated, fast, and simple operation.
  • Processes 1 to 96 samples simultaneously using a 96-well deep-well plate.
  • Also processes 1 to 24 samples simultaneously using a 24-well deep-well plate.
  • Compatible with various magnetic beads nucleic acid extraction reagents.
  • Extracts and purifies nucleic acids from human blood and body fluids.
  • Primarily used for the extraction and purification of nucleic acids in human samples.