Learn More
Nova Biomedical Corporation Auto QC Cartridge, Automated QC, Tri-Level Controls, In Vitro Diagnostic Use, Unit: 1 Each
Catalog No. NC2090071
Encompass
The Auto QC Cartridge by Nova Biomedical Corporation is an automated, three-level quality control solution designed for in vitro diagnostics. It enhances lab efficiency by reducing manual intervention through automated testing that ensures accurate blood gas and chemistry measurements. Unique features include RFID technology for tracking and seamless compatibility with the Stat Profile Prime CCS Analyzer, making it an ideal choice for laboratories prioritizing reliability and ease of use.
This item is not returnable. View return policy
Catalog No. NC2090071 Supplier Nova Biomedical Corporation Supplier No. 45150
The Auto QC Cartridge by Nova Biomedical Corporation is an automated, three-level quality control solution designed for in vitro diagnostics. It enhances lab efficiency by reducing manual intervention through automated testing that ensures accurate blood gas and chemistry measurements. Unique features include RFID technology for tracking and seamless compatibility with the Stat Profile Prime CCS Analyzer, making it an ideal choice for laboratories prioritizing reliability and ease of use.