Learn More
Thermo Scientific™ Auto-refill for VersaCool™ Refrigerated Circulating Bath
Catalog No. 13275110 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific VersaCool Refrigerated Circulating Bath
Catalog No. For Use With (Equipment)
13-275-110 Thermo Scientific VersaCool Refrigerated Circulating Bath
1 options
Catalog No. 13-275-110 Supplier Thermo Scientific™ Supplier No. 230000005
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature.
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature. Provides peace of mind with user-defined start and stop levels, as well as a time-out feature. The needle valve allows control of the fill rate from as fast a refill as possible- to a slow refill that minimizes any effect on temperature stability or any rate in between
TRUSTED_SUSTAINABILITY
Specifications
Description Auto-refill for VersaCoolandtrade Refrigerated Circulating Bath
For Use With (Equipment) Thermo Scientific VersaCool Refrigerated Circulating Bath
Type Auto-refill Device