Learn More
Malvern Panalytical Auto Sampler 2Ml Pk 100
Catalog No. NC1930576
Encompass
Autosampler Vials: 2 mL, Pk of 100+Caps. Autosampler vials. 2 mL. 100 vials and caps. These vials are also compatible with the Nanosampler for the Zetasizer Nano series. Each borosilicate glass vial includes cap and septum.
Catalog No. NC1930576 Supplier Malvern Panalytical Supplier No. BOT0036
Autosampler Vials: 2 mL, Pk of 100+Caps. Autosampler vials. 2 mL. 100 vials and caps. These vials are also compatible with the Nanosampler for the Zetasizer Nano series. Each borosilicate glass vial includes cap and septum.