Learn More
SCP Science SCP Science DigiTUBE Autosampler Tubes, Virgin Polypropylene, Ultra-Low Metal Content, 12 mL
Catalog No. NC1214533
Encompass
The SCP Science DigiTUBE Autosampler Tubes are precision-engineered from virgin polypropylene, ensuring ultra-low metal content for accurate ICP/ICP-MS analyses. Measuring 16 mm x 100 mm with a 12 mL capacity, they feature Class A graduation marks with a tolerance of +/- 0.10 mL. Their stable construction enhances chemical resistance against common acids and solvents, making them ideal for diverse lab applications. Ideal for sensitive measurements, they come in a case of 2000 tubes (2 bags of 1000).
Catalog No. NC1214533 Supplier SCP Science Supplier No. 010515629
The SCP Science DigiTUBE Autosampler Tubes are precision-engineered from virgin polypropylene, ensuring ultra-low metal content for accurate ICP/ICP-MS analyses. Measuring 16 mm x 100 mm with a 12 mL capacity, they feature Class A graduation marks with a tolerance of +/- 0.10 mL. Their stable construction enhances chemical resistance against common acids and solvents, making them ideal for diverse lab applications. Ideal for sensitive measurements, they come in a case of 2000 tubes (2 bags of 1000).