Learn More
Cliawaived Inc 10 Parameter Urinalysis Strips
SDP
Catalog No. NC1030897
Encompass_Preferred
(100/Bottle) These Urine Reagent Strips for Urinalysis are comprehensive reagent urine test strips for the detection and screening of Leukocytes, Blood, Specific Gravity, Urobilinogen, Ketone, Glucose, Protein, Bilirubin, Nitrite and pH. Urine test strips are used to monitor urine specimens for various biochemicals and their connection to certain diseases. All urine test strips yield clear and accurate results in less than 30 seconds. Results can be interpreted using visual comparison of the reagent pads to the color chart or other compatible automated Urinalysis equipment. FDA-Cleared/CLIA Waived. (FDA 510K#: K970250)
Catalog No. NC1030897 Supplier Cliawaived Inc Supplier No. CLIAU031103111
(100/Bottle) These Urine Reagent Strips for Urinalysis are comprehensive reagent urine test strips for the detection and screening of Leukocytes, Blood, Specific Gravity, Urobilinogen, Ketone, Glucose, Protein, Bilirubin, Nitrite and pH. Urine test strips are used to monitor urine specimens for various biochemicals and their connection to certain diseases. All urine test strips yield clear and accurate results in less than 30 seconds. Results can be interpreted using visual comparison of the reagent pads to the color chart or other compatible automated Urinalysis equipment. FDA-Cleared/CLIA Waived. (FDA 510K#: K970250)