Learn More
Bellco Glass, Inc. Autoblot Tube,35x225mm
SDP
Catalog No. 501946513
Encompass_Preferred
Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045. MADE IN THE USA
Catalog No. 50-194-6513 Supplier Bellco Glass, Inc. Supplier No. 791035225
Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045. MADE IN THE USA