Learn More
CPC Scientific H-Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Thr-Val-Asp-Ala-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP002A
Encompass
ONE-LETTER SEQUENCE: KKALRRQETVDALMOLECULAR FORMULA: C65H118N22O20MOLECULAR WEIGHT:1527.79STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [129198-88-5], SYNONYMS: Autocamtide-2RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: P.I. Hanson et al., Neuron., 3, 59 (1989)
Catalog No. 50-211-0156