Learn More
Enzo Life Sciences Autocamtide 2 (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1010001
Encompass_Preferred
Sequence, KKALRRQETVDAL, derived from Ca2+/calmodulin-dependent protein kinase II autophosphorylation site RQETVD plus basic residues to facilitate phosphocellulose binding. Contains consensus phosphorylation sequence R-X-X-S/T. Highly selective CaMKII substrate (Km=2 µM for CaMKII; 50 µM for PKC; no activity towards PKA). Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1224