Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Autocamtide 2, amide | 98.6% | 1526.78 | 5 MG
SDP

Catalog No. 5000422179
Encompass_Preferred

Autocamtide 2, amide is a peptide substrate (100 μM final concentration) for CaMK family assays, intended for research use only.

  • Substrate for CaMK family assays
  • Solid appearance, white to off-white color
  • Soluble in water at 50 mg/mL, requiring ultrasonic for dissolution
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Recommended storage is sealed and away from moisture

Catalog No. 50-004-22179 Supplier Medchemexpress LLC Supplier No. HYP15285MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Autocamtide 2, amide is a peptide substrate (100 μM final concentration) for CaMK family assays, intended for research use only.

  • Substrate for CaMK family assays
  • Solid appearance, white to off-white color
  • Soluble in water at 50 mg/mL, requiring ultrasonic for dissolution
  • Store powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Recommended storage is sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.