Learn More
Medchemexpress LLC HY-P0214A 1mg Medchemexpress, Autocamtide-2-related inhibitory peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0214A1MG
Encompass_Preferred
Medchemexpress, HY-P0214A 1mg Autocamtide-2-related inhibitory peptide (TFA) CAS: Autocamtide-2-related inhibitory peptide (TFA) is a highly specific and potent inhibitor of CaMKII with an IC 50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4054