Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Autocamtide-3 (acetate) | >99.9% | 1508.72 | 10 MG
SDP

Catalog No. 5000396796
Encompass_Preferred

Autocamtide-3 (acetate) is a highly pure peptide, presenting as a white to off-white solid. With a molecular weight of 1508.72, it is suitable for various research applications.

  • Purity of >99.9%
  • White to off-white solid appearance
  • Store powder sealed, away from moisture and light: -80°C for 2 years, or -20°C for 1 year
  • Store in solvent sealed, away from moisture and light: -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-003-96796 Supplier Medchemexpress LLC Supplier No. HYP3811A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Autocamtide-3 (acetate) is a highly pure peptide, presenting as a white to off-white solid. With a molecular weight of 1508.72, it is suitable for various research applications.

  • Purity of >99.9%
  • White to off-white solid appearance
  • Store powder sealed, away from moisture and light: -80°C for 2 years, or -20°C for 1 year
  • Store in solvent sealed, away from moisture and light: -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.