Learn More
Medchemexpress LLC Autocamtide-3 acetate | 99.95% | 1508.72 (free acid) | 25 MG
SDP
Catalog No. 5000396705
Encompass_Preferred
Autocamtide-3 acetate is a chemical compound supplied as a white to off-white solid with a molecular weight of 1508.72 (free acid) and a purity of 99.95% by HPLC. It requires sealed storage, away from moisture and light, at -80°C for up to two years as a powder or -80°C for six months in solvent. This product is for research use only and has not been fully validated for medical applications.
  • High purity (99.95% by HPLC)
  • White to off-white solid
  • Requires specific storage conditions for stability
  • Suitable for research applications
Catalog No. 50-003-96705 Supplier Medchemexpress LLC Supplier No. 25MGHYP3811A
Autocamtide-3 acetate is a chemical compound supplied as a white to off-white solid with a molecular weight of 1508.72 (free acid) and a purity of 99.95% by HPLC. It requires sealed storage, away from moisture and light, at -80°C for up to two years as a powder or -80°C for six months in solvent. This product is for research use only and has not been fully validated for medical applications.
  • High purity (99.95% by HPLC)
  • White to off-white solid
  • Requires specific storage conditions for stability
  • Suitable for research applications