Learn More
Fine Science Tools Instrument Marking Tape, Yellow, 290 cm
SDP
Catalog No. NC1577531
Encompass
Product: Instrument Marking Tape/ 290cm/ Tape Width: 0.6cm/ Material: Vinyl/ Color: Yellow/ Feature: Autoclave Safe
Catalog No. NC1577531 Supplier Fine Science Tools Supplier No. 2907103
Product: Instrument Marking Tape/ 290cm/ Tape Width: 0.6cm/ Material: Vinyl/ Color: Yellow/ Feature: Autoclave Safe