Learn More
Research Products International Corp Autoclavable Disposal Bags, 24 x 36 Inches, Clear, 100 per Package
SDP
Catalog No. 50998768
Encompass
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes. Made of transparent polypropylene; 0.03mm (1.5 mil) thick.
High temperature autoclavable at a maximum of 135C (275F).
Catalog No. 50-998-768 Supplier Research Products International Corp Supplier No. 246125
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes. Made of transparent polypropylene; 0.03mm (1.5 mil) thick.
High temperature autoclavable at a maximum of 135C (275F).