Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heathrow Scientific™ Autoclavable Bags

Description
- Constructed of 2.0 mm thick polypropylene.
- Can be used for repeated autoclaving.
- Strong side seams securely hold items.
- Autoclavable at 250°F (121°C).
- Front write-on panel.
- 4 inner packs of 50 bags.
- Disposable.
Specifications
Specifications
| Content And Storage | Room Temperature |
| Description | Clear Autoclavable Bags |
| Autoclavable | Autoclavable |
| Closure Type | None |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Application) | Hazardous Waste Disposal |
| For Use With (Equipment) | Autoclave |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?