Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Autoclavable Bags
GREENER_CHOICE

Catalog No. 03448269
Change view
Click to view available options
Dimensions (L x W):
310 x 660 mm
415 x 600 mm
610 x 810 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
03-448-269 610 x 810 mm
03-448-267 310 x 660 mm
03-448-268 415 x 600 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-448-269 Supplier Heathrow Scientific™ Supplier No. HS1002C
Add to Cart
Add to Cart

Clear, imprinted Autoclavable Bags with blue biohazard labeling specifically designed for safe disposal.

  • Constructed of 2.0 mm thick polypropylene.
  • Can be used for repeated autoclaving.
  • Strong side seams securely hold items.
  • Autoclavable at 250°F (121°C).
  • Front write-on panel.
  • 4 inner packs of 50 bags.
  • Disposable.

Specifications

Content And Storage Room Temperature
Description Clear Autoclavable Bags
Autoclavable Autoclavable
Closure Type None
Color Clear
Disposable Single-use
For Use With (Application) Hazardous Waste Disposal
For Use With (Equipment) Autoclave
Material Polypropylene
Sterility Non-sterile
No. per Case 200/Cs.
No. per Pack 50 Pk.
Outer Packaging Box
Thickness (English) 80 mil
Dimensions (L x W) 610 x 810 mm
Dimensions (L x W x H) 24 x 31.89 x 0.08 in.
Length (English) 24 in.
Width (English) 31.89 in.
Weight (English) 0.09 lb.
Weight (Metric) 0.04 kg
Product Type Autoclavable Bags
Capacity (English) 18.25 gal.
Capacity (Metric) 69.08 L
Length (Metric) 61 cm
Width (Metric) 81 cm
Thickness (Metric) 2 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.