Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterilis Solutions Autoclavable Biohazard Bags
SDP

Catalog No. 23777015
Change view
Click to view available options
Dimensions (L x W):
24 in. x 19 in.
24 in. x 24 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
23-777-015 24 in. x 24 in.
23-777-005 24 in. x 19 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-777-015 Supplier Sterilis Solutions Supplier No. 38232843
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Autoclavable Biohazard Bags meet all Federal and State Regulations and Requirements for Biohazard Waste

  • Autoclaving (Steam Sterilization) is the most efficient and economical wet thermal disinfection process
  • Hazardous waste producers in research and clinical laboratories rely on autoclaving to continually meet their obligated Federal and State regulations
  • OSHA required markings from Regulation 29 CFR 1910-1030 printed on each bag
  • Red Autoclavable Biohazard bags are specifically engineered in design to meet the regulations enforced worldwide
  • Compliant with the following regulations:
    • Federal DOT Regulation 49 CFR 173.197 for HazMat Handling and Markings. They exceed the 480 gram tear test as defined by ASTM D1922 in both length (MD) and width (TD) directions. They also exceed the dart drop test of 165 grams defined by ASTM D 1709 in this federal regulation.
    • EPA Resource Conservation and Recovery Act (RCRA).
    • California Health and Safety Code 117630
    • Florida Dept. Health Code # 64E-16.004-2C.  Registration #PP-2-R1-04
    • CDC Procedural Guidance on the Proper Packaging of Highly Infectious Waste
    • EU Hazardous Waste Directive
    • World Health Organization (WHO) Directive for Safe Management of Healthcare Waste for autoclaving and/or incineration

Specifications

Autoclavable Autoclavable
Closure Type None
Color Red
Disposable Reusable
For Use With (Application) Biohazardous Waste
For Use With (Equipment) Sterilis Solutions System
Material Polypropylene
Sterility Non-sterile
Quantity 100 Bags/Pk.
Temperature Range (English) 275°F
Temperature Range (Metric) 135°C
Dimensions (L x W) 24 in. x 24 in.
Capacity (English) 5 gal.
Length (Metric) 609.6 mm
Width (Metric) 609.6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.