Learn More
Electron Microscopy Sciences Autoclavable Carboy Without spigot, 20 liters
SDP
Catalog No. 501926891
Encompass_Preferred
Rugged polypropylene construction carboys are ideal for storing and dispensing large batches of double-distilled water, buffers and salts. They have ecellent chemical resistance and large handles to assist in transport. Molded-in bung accepts easy-to-use polypropylene spigot that can be operated with one hand. 83 mm polypropylene buttress screw cap includes a built-in silicone gasket for positive seal. Steam autoclavable 121 degree C (250 degree F).
FDA grade polypropylene compliant with 21CFR177.1520 for direct contact with food and beverages. USP Class VI Certified. Note: When autoclaving, do not engage cap threads.
Capacity: 20 liters
Catalog No. 50-192-6891 Supplier Electron Microscopy Sciences Supplier No. 6429916
Rugged polypropylene construction carboys are ideal for storing and dispensing large batches of double-distilled water, buffers and salts. They have ecellent chemical resistance and large handles to assist in transport. Molded-in bung accepts easy-to-use polypropylene spigot that can be operated with one hand. 83 mm polypropylene buttress screw cap includes a built-in silicone gasket for positive seal. Steam autoclavable 121 degree C (250 degree F).
FDA grade polypropylene compliant with 21CFR177.1520 for direct contact with food and beverages. USP Class VI Certified. Note: When autoclaving, do not engage cap threads.
Capacity: 20 liters