Learn More
Kuhner Shaker Inc AUTOCLAVABLE LID 121C FOR
Catalog No. 502570084
Encompass
Autoclavable lid (121°C), for sterile aeration of shake flasks with GL45 thread
Catalog No. 50-257-0084 Supplier Kuhner Shaker Inc Supplier No. SMTO00005
Autoclavable lid (121°C), for sterile aeration of shake flasks with GL45 thread