Learn More
Medicus Health Autoclavable Slide Staining Dipper with Handle
SDP
Catalog No. NC3966872
Encompass_Preferred
• Autoclavable Slide Staining Dipper with Handle, Blue holds 25 slides and is great for immunohistochemistry procedures including histological, cytological and hematological staining procedures with total immersion for slides
• Slide Stain Dipper is resistant to temperature between 0°C and 120°C
• Dippers can be sterilized by autoclave and used in the microwave
• Dipper is specially designed to grab better and allow complete closure of dish during use, avoiding reagent's evaporation
• Great for Hematology Departments who need to do Wright's Stains
Catalog No. NC3966872 Supplier Medicus Health Supplier No. 7352M4
• Autoclavable Slide Staining Dipper with Handle, Blue holds 25 slides and is great for immunohistochemistry procedures including histological, cytological and hematological staining procedures with total immersion for slides
• Slide Stain Dipper is resistant to temperature between 0°C and 120°C
• Dippers can be sterilized by autoclave and used in the microwave
• Dipper is specially designed to grab better and allow complete closure of dish during use, avoiding reagent's evaporation
• Great for Hematology Departments who need to do Wright's Stains