Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Absorbance/Fluorescence/Luminescence, 6-1536 Well, Temperature Controlled
Catalog No. 5000014389
Encompass_Preferred
The SpectraMax i3x Multi-Mode Microplate Reader is a configurable, application-upgradeable microplate reader that performs absorbance, fluorescence, and luminescence assays across a broad range of plate formats. It provides broad wavelength coverage, high sensitivity, and modular detection options for diverse biochemical and cell-based workflows.
  • Measures absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization.
  • Wavelength coverage suitable for UV-vis and fluorescence assays.
  • Xenon flash lamp and LED illumination options for flexible excitation.
  • Supports 6-1536 well plates, including standard 96 and 384 formats.
  • Top and bottom read capabilities with bottom read available for fluorescence assays.
  • Temperature control from ambient +4 °C up to 45 °C, with optional shaker and injector configurations.
  • Includes dedicated assay control and analysis software.
Catalog No. 50-000-14389 Supplier Molecular Devices Supplier No. I3X
Add to Cart
Add to Cart
Request a Quote
The SpectraMax i3x Multi-Mode Microplate Reader is a configurable, application-upgradeable microplate reader that performs absorbance, fluorescence, and luminescence assays across a broad range of plate formats. It provides broad wavelength coverage, high sensitivity, and modular detection options for diverse biochemical and cell-based workflows.
  • Measures absorbance, fluorescence, luminescence, time-resolved fluorescence, and fluorescence polarization.
  • Wavelength coverage suitable for UV-vis and fluorescence assays.
  • Xenon flash lamp and LED illumination options for flexible excitation.
  • Supports 6-1536 well plates, including standard 96 and 384 formats.
  • Top and bottom read capabilities with bottom read available for fluorescence assays.
  • Temperature control from ambient +4 °C up to 45 °C, with optional shaker and injector configurations.
  • Includes dedicated assay control and analysis software.