Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Autoclavable Waste Bags
SDP

Catalog No. 14828152 Shop All American Precision Plastics Products
Change view
Click to view available options
Capacity (English):
12 to 14 gal.
13 to 16 gal.
20 to 30 gal.
44 gal.
7 to 10 gal.
Dimensions (L x W):
19 x 24 in.
25 x 30 in.
25 x 35 in.
31 x 28 in.
31 x 38 in.
38 x 47 in.
Color:
Buff, Black Print
Clear, Red Print
Orange, Black Print
Red, Black
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Dimensions (L x W) Color
14-828-152 44 gal. 38 x 47 in. Orange, Black Print
14-828-150 44 gal. 38 x 47 in. Clear, Red Print
14-828-142 7 to 10 gal. 19 x 24 in. Red, Black
14-828-143 13 to 16 gal. 25 x 35 in. Red, Black
22-023802 12 to 14 gal. 25 x 30 in. Clear, Red Print
22-023801 7 to 10 gal. 19 x 24 in. Orange, Black Print
14-828-149 44 gal. 38 x 47 in. Buff, Black Print
14-828-147 20 to 30 gal. 31 x 38 in. Red, Black
14-828-145 20 to 30 gal. 31 x 28 in. Clear, Red Print
22-023-800 12 to 14 gal. 25 x 30 in. Red, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-828-152 Supplier Medegen Supplier No. 886
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Autoclavable Waste Bags are made of polypropylene.

  • Polypropylene
  • 1.8 mil thickness

Specifications

Capacity (Metric) 166.6 L
Capacity (English) 44 gal.
Color Orange, Black Print
Dimensions (L x W) 38 x 47 in.
Disposable Single-use
Length (Metric) 96.5 cm
Width (Metric) 119.4 cm
Autoclavable Autoclavable
Includes Indicator, flat pack, twist ties, biohazard labeling, biohazard symbol
Width (English) 47 in.
Length (English) 38 in.
Thickness 1.80 mil (0.05 mm)
Material Polypropylene
Product Type Biohazard Waste Bag
Temperature (Metric) 140.5°C
Temperature (English) 285°F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.