Learn More
Bellco Glass, Inc. AutoClave Bag,12X24"
SDP
Catalog No. 501945427
Encompass_Preferred
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving biohazrd laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 12x24"
Catalog No. 50-194-5427 Supplier Bellco Glass, Inc. Supplier No. 220012240
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving biohazrd laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 12x24"