Learn More
Bellco Glass, Inc. AutoClave Bag,24X30"
SDP
Catalog No. 501945428
Encompass_Preferred
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving biohazrd laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 24x30"
Catalog No. 50-194-5428 Supplier Bellco Glass, Inc. Supplier No. 220024300
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving biohazrd laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 24x30"