Learn More
Bellco Glass, Inc. Autoclave Bag,24X36"
SDP
Catalog No. 501945429
Encompass_Preferred
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving bio-hazard laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 24x36"
Catalog No. 50-194-5429 Supplier Bellco Glass, Inc. Supplier No. 220024360
Rugged 50ml polypropylene bags with OSHA bio-hazard symbol imprinted on the bags for autoclaving bio-hazard laboratory waste.. MADE IN THE USA Temperature rating to 130 degrees c Size: 24x36"