Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Autoclave Bag

Catalog No. 22043043 Shop All Globe Scientific Products
Change view
Click to view available options
Dimensions (L x W):
8 x 12 in.
12 x 20 in.
24 x 32 in.
Color:
Clear
Red
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
22-043-043 24 x 32 in. Red
22-043-038 8 x 12 in. Clear
22-043-039 8 x 12 in. Red
22-043-040 12 x 20 in. Clear
22-043-041 12 x 20 in. Red
22-043-042 24 x 32 in. Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-043-043 Supplier Globe Scientific Supplier No. 4854R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for the safe collection, containment, and sterilization of laboratory biohazardous waste and contaminated utensils

  • For the disposal of containers, tubes, pipette tips, culture plates, PPE, gloves, and more, making them ideal for use in clinical and research labs, hospitals, physicians offi ces and wherever hazardous waste is a concern
  • Eliminate the high cost while minimizing regulatory and environmental concerns associated with hazardous waste disposal
  • Features:
    • 2 mm gauge
    • Imprinted biohazard symbol
    • Available in clear or red
    • Clear bags autoclavable up to 134°C/273.2°F
    • Red bags autoclavable up to 121°C/249.8°F
  • Material:
    • Clear bags: Polypropylene (PP)
    • Red bags: High-Density Polyethylene (HDPE)

Specifications

Autoclavable Autoclavable
Closure Type None
Color Red
Disposable Single-use
Material HDPE
Sterility Non-sterile
No. per Case 4 Pk./Cs.
No. per Pack 50/Pk.
Dimensions (L x W) 24 x 32 in.
Length (English) 24 in.
Width (English) 32 in.
Product Type Autoclave Bags
Length (Metric) 60.96 cm
Width (Metric) 81.28 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.