Learn More
Research Products International Corp Autoclave Bag Clips, 100 per Package
SDP
Catalog No. 50998769
Encompass
Unique designed clip keeps bag securely closed during autoclaving yet still allows the bag to release internal pressure that can build up during atuoclaving. Clip opens and closes easily using one hand. Clip remains on bag for final disposal.
Fits bag sizes from 12 x 24 inches up to 19 x 23 inches.
Catalog No. 50-998-769 Supplier Research Products International Corp Supplier No. 246141
Unique designed clip keeps bag securely closed during autoclaving yet still allows the bag to release internal pressure that can build up during atuoclaving. Clip opens and closes easily using one hand. Clip remains on bag for final disposal.
Fits bag sizes from 12 x 24 inches up to 19 x 23 inches.