Learn More
JDM LAB SOLUTIONS LLC Autoclave bag, drawstring, red, 33 x 41"
Catalog No. NC4037092
Encompass
Bowtie™ Biohazard bags, autoclave bag with drawstring, PE, 33 x 41in. (84 x 104cm), with marking area and sterilization indicator, 100/pk
Catalog No. NC4037092 Supplier JDM LAB SOLUTIONS LLC Supplier No. JDMA8003R
Bowtie™ Biohazard bags, autoclave bag with drawstring, PE, 33 x 41in. (84 x 104cm), with marking area and sterilization indicator, 100/pk