Learn More
MTC Bio Bowtie™ Biohazard bags, autoclave bag with drawstring, PE, 25 x 35in. (64 x 89cm), with marking area and sterilization indicator, 100/pk
Catalog No. NC2017794
Encompass_Preferred
MTC Bio’s new BowTie™ Biohazard Bags apply a modern touch to a basic lab necessity that hasn’t been improved in decades. These clearly marked bags are equiped with an exclusive easy-pull drawstring. They also include a sterilization indicator area that changes color after autoclaving, for foolproof verification of sterility. Bags are clearly marked with biohazard symbols and includes a large white marking area for custom identification. BowTie™ Bags are made of HDPE and are suitable for autoclaving at 121°C cycles.
Catalog No. NC2017794 Supplier MTC Bio Supplier No. A8001R
MTC Bio’s new BowTie™ Biohazard Bags apply a modern touch to a basic lab necessity that hasn’t been improved in decades. These clearly marked bags are equiped with an exclusive easy-pull drawstring. They also include a sterilization indicator area that changes color after autoclaving, for foolproof verification of sterility. Bags are clearly marked with biohazard symbols and includes a large white marking area for custom identification. BowTie™ Bags are made of HDPE and are suitable for autoclaving at 121°C cycles.