Learn More
MTC Bio Autoclave bags, 10" x 12.5" ( 25cmx32cm) red, biohazard, printed, marking area, box of 100
Catalog No. NC3345648
Encompass_Preferred
These high temperature autoclavable bags are manufactured from heavy gauge polypropylene material that is not only more puncture-resistant than polyethylene, but also withstands autoclaving at temperatures up to 134˚C (270˚F). Their ability to contain is completely unaffected at standard autoclave temperature (121˚C).Each bag has an oversized white marking panel, suitable for use with all types of permanent markers. They are also prominently marked with the words “Hazardous Waste” and the standard “Biohazard” symbol, conforming to the OSHA standard (29CFR1910.1030).
Catalog No. NC3345648 Supplier MTC Bio Supplier No. A9006R
These high temperature autoclavable bags are manufactured from heavy gauge polypropylene material that is not only more puncture-resistant than polyethylene, but also withstands autoclaving at temperatures up to 134˚C (270˚F). Their ability to contain is completely unaffected at standard autoclave temperature (121˚C).Each bag has an oversized white marking panel, suitable for use with all types of permanent markers. They are also prominently marked with the words “Hazardous Waste” and the standard “Biohazard” symbol, conforming to the OSHA standard (29CFR1910.1030).