Learn More
Research Products International Corp Autoclave Bags, 12 x 24 Inches, 1 1/4 mil Thick, 100 per Package
SDP
Catalog No. 50476661
Encompass
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 100 per package.
Catalog No. 50-476-661 Supplier Research Products International Corp Supplier No. 248253
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 100 per package.