Learn More
Research Products International Corp Autoclave Bags, 30 x 34 Inches, 2 mil Thick, 100 per Package
SDP
Catalog No. 50212565
Encompass
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Bags allow full visibility of medical supplies and supplies. Supplied 100 per package.
Catalog No. 50-212-565 Supplier Research Products International Corp Supplier No. 248255
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Bags allow full visibility of medical supplies and supplies. Supplied 100 per package.