Learn More
Research Products International Corp Autoclave Basket, 4 3/4 x 4 1/4 x 5 1/2 Inches, 6 per Package
SDP
Catalog No. 50212578
Encompass
Ideal for rinsing or draining glassware. Works great for moving small objects in and out of an autoclave. Can also be used for storage of test tubes or culture tubes inside incubators. All polypropylene construction will not corrode or stain metal items or glassware.
Catalog No. 50-212-578 Supplier Research Products International Corp Supplier No. 248644
Ideal for rinsing or draining glassware. Works great for moving small objects in and out of an autoclave. Can also be used for storage of test tubes or culture tubes inside incubators. All polypropylene construction will not corrode or stain metal items or glassware.